Learn More
H3K9me2S10phK27me2S28ph Monoclonal Antibody (51TA-2H12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA3065
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA3-065 detects dimethyl-phospho-histone H3 Lys9/Lys27 pSer10/pSer28 in human samples and has successfully been used in western blot and immunoprecipitation applications. The MA3-065 immunogen is a synthetic peptide conjugated to ovalbumin corresponding to residues 180-195 of human TAF10 and only recognizes phosphorlayted pSer10 or pSer28 of Histone H3 when the adjacent Lys is dimethylated.
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.Spezifikation
H3K9me2S10phK27me2S28ph | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.05% sodium azide | |
P68431, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7 | |
Ovalbumin conjugated synthetic peptide (ASGSSRSKS[dimethyl-K]DRKYTLC) correspeonding to aa 180-195 of human TAF10 | |
RUO | |
126961, 333932, 8350, 8356, 8357, 8358, 8968 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Immunoprecipitation, Western Blot | |
51TA-2H12 | |
Unconjugated | |
H3C1 | |
Di histone H3/c; dimethyl H3; di-methyl H3; Dimethyl Histone H3; di-methyl histone H3; Di-Methyl-Phospho-H3 K9 K27 S10 S28; Di-Methyl-Phospho-H3 Lys9 Lys27 Ser10 Ser28; Di-Methyl-Phospho-Histone H3 K9 K27 S10 S28; Di-Methyl-Phospho-Histone H3 Lys9 Lys27 Ser10 Ser28; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3C; H3 K9me2S10phK27me2S28ph; H3.2; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-3A; H3-3B; H3a; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3F1K; H3F2; H3F3; H3F3A; H3F3B; H3F3C; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; H3k27; H3K27me2; H3K9; H3K9me2; H3K9me3S10phK27me3S28ph; H3K9me3S10pK27me3S28p; H3Lys9me3Ser10phLys27me3Ser28ph; H3Lys9me3Ser10pL | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |