Learn More
Homosalate, MedChemExpress
Artikelnummer. 16488516
Encompass_Preferred
Change view
Click to view available options
Menge:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
16488516 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16488516 Lieferant Medchem Express Lieferanten-Nr. HYB0928
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
MedChemExpress Homosalate is an organic compound used in some sunscreens, it is used as a chemical UV filter, protecting the skin from sun damage.
Homosalate is an organic compound used in some sunscreens, it is used as a chemical UV filter, protecting the skin from sun damage.
Spezifikation
Chemischer Name oder Material Homosalate
CAS 118-56-9
Farbe Colorless
Hinweise zur Reinheitsqualität Research
Summenformel C16H22O3
Menge 100 mg
Synonym Homomenthyl salicylate
Löslichkeitsinformationen DMSO : 100 mg/mL (381.18 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(OC1CC(C)(C)CC(C)C1)C2=CC=CC=C2O
Molekulargewicht (g/mol) 262.34
Formelmasse 262.34
Reinheit (%) 95.0%
Güte Research
Physikalische Form Liquid