Learn More
HPV-16 Mouse, Clone: HPV16/1296, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio MSM4-1296-P0
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| HPV-16 | |
| Nuclear | |
| HPV16/1296 | |
| Unconjugated | |
| Mouse | |
| HPV-16 infected cells or cervical tissue | |
| 20 μg | |
| Primary | |
| IgG2a κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| Human papilloma virus type 16 | |
| Protein A/G Purified | |
| RUO | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |