missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD11B1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518331
Weitere Details : Netto-Gewicht : 0.01000kg
Description
This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
HSD11B1 is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, HSD11B1 can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene.Specifications
HSD11B1 | |
Polyclonal | |
Unconjugated | |
Hsd11b1 | |
11 beta-hydroxysteroid dehydrogenase type 1; 11beta-HSD type 1; 11-beta-HSD1; 11beta-HSD1; 11beta-HSD-1; 11beta-HSD1A; 11-beta-hydroxysteroid dehydrogenase; 11-beta-hydroxysteroid dehydrogenase 1; 11-beta-hydroxysteroid dehydrogenase type 1; 11beta-hydroxysteroid dehydrogenase type 1; 11-DH; Corticosteroid 11-beta-dehydrogenase isozyme 1; Corticosteroid 11-beta-dehydrogenase, isozyme 1 (11-DH) (11-beta-hydroxysteroid dehydrogenase 1) (11-beta-HSD1) (11beta-HSD1A); cortocosteroid 11-beta dehydrogenase; CORTRD2; HDL; HSD11; HSD11B; Hsd11b1; HSD11L; hydroxysteroid (11-beta) dehydrogenase 1; hydroxysteroid 11-beta dehydrogenase 1; hydroxysteroid dehydrogenase, 11 beta type 1; LRRGT00065; MGC13539; OTTHUMP00000034650; SDR26C1; short chain dehydrogenase/reductase family 26C member 1; short chain dehydrogenase/reductase family 26C, member 1; short-chain reductase; short-chain reductase/dehydrogenase | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
3290 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P28845 | |
Hsd11b1 | |
Synthetic peptide sequence (CTSYNMDRFINK) corresponding to the C-terminus amino acids of HSD11B1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |