missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD3B2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527791
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
3 beta-hydroxysteroid dehydrogenase (3beta-HSD), also known as HSD3B1 or HSDB3, is a bifunctional enzyme that plays a crucial role in the synthesis of all classes of hormonal steroids. Two human 3beta-HSD proteins, designated type I (3beta-HSD) and type II (3beta-HSD2), are expressed by different genes and function in different areas of the body. Localized to the membrane of the endoplasmic reticulum (ER) and expressed in skin and placenta, 3beta-HSD is the type I protein that catalyzes the oxidative conversion of delta5-ene-3-beta-hydroxy steroid, as well as the conversion of various ketosteroids. Defects in the gene encoding 3beta-HSD are associated with classic salt wasting, genital ambiguity, hypogonadism, insulin-resistant polycystic ovary syndrome (PCOS) and an increased susceptibility to prostate cancer. Additionally, congenital deficiency of 3beta-HSD activity results in a severe depletion of steroid formation which can be lethal in young children.Spezifikation
HSD3B2 | |
Polyclonal | |
Unconjugated | |
HSD3B2 | |
3 beta HSD II; 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase 2; 3 beta-hydroxysteroid dehydrogenase 6; 3 beta-hydroxysteroid dehydrogenase type II, delta 5-delta 4-isomerase type II, 3 beta-HSD type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 4; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II; 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type IV; 3beta-HSD; 3-beta-HSD adrenal and gonadal type; 3-beta-HSD II; 3-beta-HSD IV; 3-beta-hydroxy-5-ene steroid dehydrogenase; 3-beta-hydroxy-Delta(5)-steroid dehydrogenase; delta 5-delta 4-isomerase type II; Delta-5-3-ketosteroid isomerase; HSD3B; Hsd3b1; Hsd3b2; Hsd3b6; HSD-3beta; HSDB; HSDB3B; Hydroxy-delta-5-steroid dehydrogenase 3 beta- and steroid delta-isomerase; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2; hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15493, 3284 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.24 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P26149, P26439 | |
HSD3B2 | |
Recombinant fragment corresponding to a region within amino acids 126 and 372 of Human HSD3B2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |