missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP40 Monoclonal Antibody (G.779.5), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA514869
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other HSPs.
HSP40 Monoclonal antibody specifically detects HSP40 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HSP40 | |
Monoclonal | |
Unconjugated | |
DNAJB1 | |
0610007I11Rik; DjB1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ heat shock protein family (Hsp40) member B1; dnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJ1; Dnajb1; HDJ1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; HSPF1; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; RSPH16B; Sis1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3337, 361384, 81489 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
G.779.5 | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P25685, Q9QYJ3 | |
DNAJB1 | |
Synthetic peptide corresponding to Glu223 of human HSP40/Hdj1 | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |