Learn More
HTR5A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532867
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Elephant, Panda, Horse, Guinea pig (100%); Opossum (94%); Turkey, Chicken (89%); Lizard, Zebrafish, Stickleback (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
The neurotransmitter serotonin (5-hydroxytryptamine, 5-HT) has been implicated in a wide range of psychiatric conditions and also has vasoconstrictive and vasodilatory effects. The gene described in this record is a member of 5-hydroxytryptamine (serotonin) receptor family and encodes a multi-pass membrane protein that functions as a receptor for 5-hydroxytryptamine and couples to G-proteins. This protein has been shown to function in part through the regulation of intracellular Ca2+ mobilization.Spezifikation
HTR5A | |
Polyclonal | |
Unconjugated | |
HTR5A | |
5-HT-5; 5HT5A; 5-HT5A; 5-HT-5A; 5-HT5a receptor; 5-hydroxytryptamine (serotonin) receptor 5A; 5-hydroxytryptamine (serotonin) receptor 5A, G protein-coupled; 5-hydroxytryptamine receptor 5A; 5-hydroxytryptamine5a; Htr5; Htr5a; MR22; REC17; Serotonin receptor 5A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100063709, 15563, 25689, 3361, 482814, 541200 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P30966, P35364, P47898 | |
HTR5A | |
Synthetic 18 amino acid peptide from 1st extracellular domain of human 5HT5A Receptor. | |
50 μg | |
Primary | |
Bovine, Canine, Equine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |