Learn More
Abnova™ Human ADPRH Full-length ORF (NP_001116.1, 1 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000141-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The enzyme encoded by this gene catalyzes removal of mono-ADP-ribose from arginine residues of proteins in the ADP-ribosylation cycle. Unlike the rat and mouse enzymes, which require DTT for maximal activity, the human enzyme is DTT-independent. [provided by RefSeq]
Sequence: MEKYVAAMVLSAAGDALGYYNGKWEFLQDGEKIHRQLAQLGGLDALDVGRWRVSDDTVMHLATAEALVEAGKAPKLTQLYYLLAKHYQDCMEDMDGRAPGGASVHNAMQLKPGKPNGWRIPFNSHEGGCGAAMRAMCIGLRFPHHSQLDTLIQVSIESGRMTHHHPTGYLGALASALFTAYAVNSRPPLQWGKGLMELLPEAKKYIVQSGYFVEENLQHWSYFQTKWENYLKLRGILDGESAPTFPESFGVKERDQFYTSLSYSGWGGSSGHDAPMIAYDAVLAAGDSWKELAHRAFFHGGDSDSTAAIAGCWWGVMYGFKGVSPSNYEKLEYRNRLEETARALYSLGSKEDTVISLSpezifikation
NP_001116.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ADPRH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
141 | |
ADPRH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARH1 | |
ADPRH | |
Recombinant | |
wheat germ expression system |