Learn More
ACROBiosystems Human ALK-1 / ACVRL1 Protein, His Tag
Marke:  ACROBiosystems AL1-H5227-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15895897
  • 2258.12 € / 1 Milligramm
Verfügbar ab: 02-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Serine/threonine-protein kinase receptor R3 is an enzyme that in humans is encoded by the ALK1 gene. ALK1 is a receptor in the TGF beta signaling pathway. ALK1 protein is a receptor in the TGF beta signaling pathway. It plays an important role in vascular development, remodeling, and pathologic angiogenesis, play a role in stabilizing angiogenic vessels and contribute to resistance to anti-VEGF therapies, ALK1 blockade may represent an effective therapeutic opportunity complementary to the current antiangiogenic modalities in the clinic. Recently, researcher found that, ALK1-Fc inhibited BMP9-mediated Id-1 expression in human umbilical vein endothelial cells and inhibited cord formation by these cells on a Matrigel substrate, in a chick chorioallantoic membrane assay, ALK1-Fc reduced vascular endothelial growth factor-, fibroblast growth factor-, and BMP10-mediated vessel formation, and ALK1-Fc treatment reduced tumor burden in mice receiving orthotopic grafts of MCF7 mammary adenocarcinoma cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.5 kDa. The protein migrates as 22-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_000011.2
11.5 kDa
1 mg
Asp 22 - Gln 118
ACVRL1,ACVRLK1,ALK-1,HHT,HHT2,ORW2,SKR3,TSR-I
Unconjugated
PBS, pH7.4
HEK293
Human
Powder
>97% as determined by SDS-PAGE
Human ALK-1, His Tag (AL1-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Gln 118 (Accession # NP_000011.2).
-20°C
ALK-1
His Tag
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens