Learn More
Abnova™ Human AMPD3 Partial ORF (NP_000471, 2 a.a. - 82 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000272-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the AMP deaminase gene family. The encoded protein is a highly regulated enzyme that catalyzes the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. This gene encodes the erythrocyte (E) isoforms, whereas other family members encode isoforms that predominate in muscle (M) and liver (L) cells. Mutations in this gene lead to the clinically asymptomatic, autosomal recessive condition erythrocyte AMP deaminase deficiency. Alternatively spliced transcript variants encoding different isoforms of this gene have been described. [provided by RefSeq]
Sequence: ALSSEPAEMPRQFPKLNISEVDEQVRLLAEKVFAKVLREEDSKDALSLFTVPEDCPIGQKEAKERELQKELAEQKSVETAKSpezifikation
NP_000471 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.65kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ALSSEPAEMPRQFPKLNISEVDEQVRLLAEKVFAKVLREEDSKDALSLFTVPEDCPIGQKEAKERELQKELAEQKSVETAK | |
RUO | |
AMPD3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
272 | |
AMPD3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMPD3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |