missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARHGEF4 Partial ORF (NP_056135, 581 a.a. - 690 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00050649-Q01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16166406

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 07-06-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. This protein is similar to rat collybistin protein. Alternative splicing of this gene generates two transcript variants which encode different isoforms. Also there is possibility for the usage of multiple polyadenylation sites for this gene. [provided by RefSeq]

Sequence: GDSHLLCTRKPEQKQRWLKAFAREREQVQLDQETGFSITELQRKQAMLNASKQQVTGKPKAVGRPCYLTRQKHPALPSNRPQQQVLVLAEPRRKPSTFWHSISRLAPFRK
Spezifikation

Spezifikation

NP_056135
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.95kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GDSHLLCTRKPEQKQRWLKAFAREREQVQLDQETGFSITELQRKQAMLNASKQQVTGKPKAVGRPCYLTRQKHPALPSNRPQQQVLVLAEPRRKPSTFWHSISRLAPFRK
RUO
ARHGEF4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
50649
ARHGEF4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASEF/ASEF1/GEF4/STM6
ARHGEF4
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote