Learn More
Abnova™ Human ATP6V1G3 Full-length ORF (NP_573569.1, 1 a.a. - 118 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00127124-P01.10ug
Beschreibung
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″ and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene encodes one of three G subunit proteins. Transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MTSQSQGIHQLLQAEKRAKDKLEEAKKRKGKRLKQAKEEAMVEIDQYRMQRDKEFRLKQSKIMGSQNNLSDEIEEQTLGKIQELNGHYNKYMESVMNQLLSMVCDMKPEIHVNYRATNSpezifikation
NP_573569.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ATP6V1G3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
127124 | |
ATP6V1G3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP6G3/MGC119810/MGC119813/Vma10 | |
ATP6V1G3 | |
Recombinant | |
wheat germ expression system |