Learn More
ACROBiosystems Human B7-H3 (4Ig) / B7-H3b Protein, Fc Tag
Marke: ACROBiosystems B7B-H5258-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Human B7 homolog 3 (B7-H3) is a member of the B7 family of immune proteins that provide signals for the regulation of immune responses. Other family members include B7-1, B7-2, B7-H1/PD-L1, B7-H2, and PD-L2. B7 family proteins are type I transmembrane immunoglobulin (Ig) superfamily members that contain extracellular Ig V‐like and Ig C‐like domains with a short cytoplasmic tail. Termed 4IgB7-H3 or B7-H3b, this molecule has two additional Ig-like domains (one V‐type and one C‐type) and shows a ubiquituous expression pattern.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.1 kDa. The protein migrates as 100-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001019907.1 | |
1 mg | |
Gly 27 - Thr 461 | |
4 Ig-B7-H3,B7-H3,CD276,PSEC0249,UNQ309/PRO352,B7 homolog 3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |
73.1 kDa | |
Human B7-H3 (4 Ig), Fc Tag (B7B-H5258) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (Accession # Q5ZPR3-1). | |
-20°C | |
B7-H3 (4 Ig) | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |