Learn More
ACROBiosystems Human B7-H5 / Gi24 / VISTA Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marke:  ACROBiosystems B75-H5258-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15895967
  • 2661.24 € / 1 Milligramm
Verfügbar ab: 03-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Platelet receptor Gi24, also known as B7-H5 and stress-induced secreted protein-1 (Sisp-1), is a protein that in humans is encoded by the C10orf54 gene, which contains 1 Ig-like (immunoglobulin-like) domain. As for C10orf54 gene, C10orf54 appears to positively interact with BMP-4, potentiating BMP signaling and the transition from an undifferentiated to a differentiated state on ESCs. Human C10orf54 undergoes proteolytic cleavage by MT1-MMP, generating a soluble 30 kDa extracellular fragment plus a 25-30 kDa membrane-bound fragment.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH20568.1
1 mg
Phe 33 - Ala 194
B7-H5,SISP1,Gi24,VISTA
Unconjugated
Mouse IgG Fc Tag
Less than 0.1 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
45.0 kDa
Human B7-H5, Mouse IgG2a Fc Tag, low endotoxin (B75-H5258) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568.1).
-20°C
B7-H5
Human
50 mM Tris, 100 mM Glycine, pH7.5
HEK293
Immune Checkpoint Proteins