Learn More
Abnova™ Human BAP1 Full-length ORF (NP_004647.1, 1 a.a. - 729 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008314-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene localizes to the nucleus and it interacts with the RING finger domain of the breast cancer 1, early onset protein (BRCA1). This gene is thought to be a tumor suppressor gene that functions in the BRCA1 growth control pathway. There are multiple polyadenylation sites found in this gene. [provided by RefSeq]
Sequence: MNKGWLELESDPGLFTLLVEDFGVKGVQVEEIYDLQSKCQGPVYGFIFLFKWIEERRSRRKVSTLVDDTSVIDDDIVNNMFFAHQLIPNSCATHALLSVLLNCSSVDLGPTLSRMKDFTKGFSPESKGYAIGNAPELAKAHNSHARPEPRHLPEKQNGLSAVRTMEAFHFVSYVPITGRLFELDGLKVYPIDHGPWGEDEEWTDKARRVIMERIGLATAGEPYHDIRFNLMAVVPDRRIKYEARLHVLKVNRQTVLEALQQLIRVTQPELIQTHKSQESQLPEESKSASNKSPLVLEANRAPAASEGNHTDGAEEAAGSCAQAPSHSPPNKPKLVVKPPGSSLNGVHPNPTPIVQRLPAFLDNHNYAKSPMQEEEDLAAGVGRSRVPVRPPQQYSDDEDDYEDDEEDDVQNTNSALRYKGKGTGKPGALSGSADGQLSVLQPNTINVLAEKLKESQKDLSIPLSIKTSSGAGSPAVAVPTHSQPSPTPSNESTDTASEIGSAFNSPLRSPIRSANPTRPSSPVTSHISKVLFGEDDSLLRVDCIRYNRAVRDLGPVISTGLLHLAEDGVLSPLALTEGGKGSSPSIRPIQGSQGSSSPVEKEVVEATDSREKTGMVRPGEPLSGEKYSPKELLALLKCVEAEIANYEACLKEEVEKRKKFKIDDQRRTHNYDEFICTFISMLAQEGMLANLVEQNISVRRRQGVSIGRLHKQRKPDRRKRSRPYKAKRQSpezifikation
NP_004647.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
106.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
BAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8314 | |
BAP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686N04275/FLJ35406/FLJ37180/HUCEP-13/KIAA0272/UCHL2/hucep-6 | |
BAP1 | |
Recombinant | |
wheat germ expression system |