Learn More
ACROBiosystems Human BCMA / TNFRSF17 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marke:  ACROBiosystems BCA-H5253-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16407219
  • 3108.82 € / 1 Milligramm
Verfügbar ab: 17-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 32.8 kDa. The protein migrates as 35-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_001183.2
1 mg
Met 1 - Ala 54
TNFRSF17,CD269,BCM,BCMA
Unconjugated
Mouse IgG Fc Tag
Less than 0.1 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
32.8 kDa
Human BCMA, Mouse IgG2a Fc Tag (BCA-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).
-20°C
BCMA
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens