missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human BMPR-IA / ALK-3 Protein, Fc Tag
290.32 € - 1100.79 €
Spezifikation
Zugriffsnummer | CAA80257 |
---|---|
Molekulargewicht | 40.3 kDa |
Quelle | Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession # P36894-1). |
Immunogen | Gln 24 - Arg 152 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15835907
|
ACROBiosystems
AL3-H5259-200UG |
200 μg |
290.32 €
200 Mikrogramm |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15825907
|
ACROBiosystems
AL3-H5259-1MG |
1 mg |
1100.79 €
1 Milligramm |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The bone morphogenetic protein receptor, type IA is also known as BMPR1A or ALK3 is a protein which in humans is encoded by the BMPR1A gene. BMPR1A has also been designated as CD292 (cluster of differentiation 290). The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A (this protein) and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2[1]. Alk3-mediated BMP signaling in AV endocardial/mesenchymal cells plays a central role during cushion morphogenesis.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.3 kDa. The protein migrates as 56 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
CAA80257 | |
Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession # P36894-1). | |
-20°C | |
BMPR-IA | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
40.3 kDa | |
Gln 24 - Arg 152 | |
BMPR-1 A,ALK-3,10q23del,ACVRLK3,CD292,SKR5 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |