Learn More
ACROBiosystems Human CD30 / TNFRSF8 Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
Marke:  ACROBiosystems TN8-H5250-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15887827
  • 2634.48 € / 1 Milligramm
Verfügbar ab: 23-12-2022
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Human CD30 is also known as TNFRSF8, is a cell membrane protein of the tumor necrosis factor receptor family and tumor marker. TNFRSF-8 is expressed by activated, but not by resting, T and B cells. Also, CD30 is expressed on classical Hodgkin Lymphoma cells together with CD15. CD30 is the receptor for TNFSF8/CD30L. CD30 can interact with TRAF2 and TRAF5, and mediate the signal transduction that leads to the activation of NF-kappa-B. TNFRSF8 may play a role in the regulation of cellular growth and transformation of activated lymphoblasts. TNFRSF8 is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity.
This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 66.3 kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_001234.2
66.3 kDa
1 mg
Phe 19 - Lys 379
TNFRSF8,CD30,D1S166E,Ki-1
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Human CD30, Llama IgG2b Fc Tag, low endotoxin (TN8-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).
-20°C
CD30
Llama IgG Fc Tag
Less than 0.01 EU / µg by the LAL method.
Immune Checkpoint Proteins