Learn More
Abnova™ Human CD300LB Full-length ORF (NP_777552.1, 1 a.a. - 238 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00124599-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD300LB is a nonclassical activating receptor of the immunoglobulin (Ig) superfamily expressed on myeloid cells (Martinez-Barriocanal and Sayos, 2006 [PubMed 16920917]).[supplied by OMIM]
Sequence: MCRRCKPELGQNFQSASGICICHWLQIRRTRSREGRAMWLPPALLLLSLSGCFSIQGPESVRAPEQGSLTVQCHYKQGWETYIKWWCRGVRWDTCKILIETRGSEQGEKSDRVSIKDNQKDRTFTVTMEGLRRDDADVYWCGIERRGPDLGTQVKVIVDPEGAASTTASSPTNSNMAVFIGSHKRNHYMLLVFVKVPILLILVTAILWLKGSQRVPEEPGEQPIYMNFSEPLTKDMATSpezifikation
NP_777552.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD300b/CLM7/IREM3/TREM5 | |
CD300LB | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
124599 | |
CD300LB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCRRCKPELGQNFQSASGICICHWLQIRRTRSREGRAMWLPPALLLLSLSGCFSIQGPESVRAPEQGSLTVQCHYKQGWETYIKWWCRGVRWDTCKILIETRGSEQGEKSDRVSIKDNQKDRTFTVTMEGLRRDDADVYWCGIERRGPDLGTQVKVIVDPEGAASTTASSPTNSNMAVFIGSHKRNHYMLLVFVKVPILLILVTAILWLKGSQRVPEEPGEQPIYMNFSEPLTKDMAT | |
RUO | |
CD300LB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |