Learn More
ACROBiosystems Human CD38 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
423.38 € - 2951.55 €
Spezifikation
Zugriffsnummer | NP_001766.2 |
---|---|
Molekulargewicht | 56.9 kDa |
Quelle | Human CD38, mouse IgG2a Fc Tag, low endotoxin (CD8-H5253) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # P28907-1). |
Immunogen | Val 43 - Ile 300 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15886297
|
ACROBiosystems
CD8-H5253-1MG |
1 mg |
2951.55 €
1 Milligramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15896297
|
ACROBiosystems
CD8-H5253-50UG |
50 μg |
423.38 €
50 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
CD antigen CD38 is also known as ADP-ribosyl cyclase 1, which belongs to the ADP-ribosyl cyclase family. CD38 is expressed at high levels in pancreas, liver, kidney, brain, testis, ovary, placenta, malignant lymphoma and neuroblastoma. CD38 is a multifunctional ectoenzyme that catalyzes the synthesis and hydrolysis of cyclic ADP-ribose (cADPR) from NAD+ to ADP-ribose. These reaction products are essential for the regulation of intracellular Ca2+. The loss of CD38 function is associated with impaired immune responses, metabolic disturbances, and behavioral modifications. The CD38 protein is a marker of cell activation. It has been connected to HIV infection, leukemias, myelomas, solid tumors, type II diabetes mellitus and bone metabolism. CD38 has been used as a prognostic marker in leukemia.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 56.9 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001766.2 | |
Human CD38, mouse IgG2a Fc Tag, low endotoxin (CD8-H5253) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # P28907-1). | |
-20°C | |
CD38 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
56.9 kDa | |
Val 43 - Ile 300 | |
CD38,T10,cADPr hydrolase 1 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |