Learn More
ACROBiosystems Human CD40 / TNFRSF5 Protein, His Tag
MALS verified
Marke: ACROBiosystems CD0-H5228-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 5 (TNFRSF5) is also known as CD40, is a member of the TNF receptor superfamily. The expression of CD40 is diverse. TNFRSF5 has been found to be essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. CD40 is the receptor for TNFSF5/CD40LG. Defects in CD40 are the cause of immunodeficiency with hyper-IgM type 3 (HIGM3).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 29-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH12419.1 | |
1 mg | |
Glu 21 - Arg 193 | |
CD40,Bp50,CDW40,MGC9013,TNFRSF5,p50 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
21.1 kDa | |
Human CD40, His Tag (MALS verified) (CD0-H5228) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1). | |
-20°C | |
CD40 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |