Learn More
ACROBiosystems Human CD69 / CLEC2C Protein, His Tag
338.70 € - 2153.18 €
Spezifikation
Zugriffsnummer | AAH07037 |
---|---|
Molekulargewicht | 16.8 kDa |
Quelle | Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). It contains AA Ser 62 - Lys 199 (Accession # AAH07037). |
Immunogen | Ser 62 - Lys 199 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15886317
|
ACROBiosystems
CD9-H5222-100UG |
100 μg |
338.70 €
100 Mikrogramm |
Verfügbar ab: 06-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15896317
|
ACROBiosystems
CD9-H5222-1MG |
1 mg |
2153.18 €
1 Milligramm |
Verfügbar ab: 06-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Early activation antigen CD69 is also known as C-type lectin domain family 2 member C (CLEC2C), Activation inducer molecule (AIM), Early T-cell activation antigen p60, Leukocyte surface antigen Leu-23. CD69 contains one C-type lectin domain. CD69 / CLEC2C is expressed on the surface of activated T-cells, B-cells, natural killer cells, neutrophils, eosinophils, epidermal Langerhans cells and platelets. CD69 is induced by antigens, mitogens or activators of PKC on the surface of T and B-lymphocytes and by interaction of IL-2 with the p75 IL-2R on the surface of NK cells. CD69 / CLEC2C involved in lymphocyte proliferation and functions as a signal transmitting receptor in lymphocytes, natural killer (NK) cells, and platelets.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 20-26 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Spezifikation
AAH07037 | |
Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). It contains AA Ser 62 - Lys 199 (Accession # AAH07037). | |
-20°C | |
CD69 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
16.8 kDa | |
Ser 62 - Lys 199 | |
CD69,CLEC2C,AIM,BL-AC/P26,EA1,GP32/28,MLR-3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |