missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human CD7 Protein, His Tag
447.57 € - 3253.97 €
Spezifikation
Zugriffsnummer | NP_006128.1 |
---|---|
Molekulargewicht | 18.3 kDa |
Quelle | Human CD7, His Tag (CD7-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1). |
Immunogen | Ala 26 - Pro 180 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17116141
|
ACROBiosystems
CD7-H52H7-1MG |
1 mg |
3253.97 €
1 Milligramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
17106141
|
ACROBiosystems
CD7-H52H7-100UG |
100 μg |
447.57 €
100 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
T-cell antigen CD7 (CD7) is also known as GP40, LEU-9, TP41 and Tp40. CD7 is a protein that in humans is encoded by the CD7 gene, this gene encodes a transmembrane protein which is a member of the immunoglobulin superfamily. CD7 has been shown to interact with PIK3R1. This protein is found on thymocytes and mature T cells. It plays an essential role in T-cell interactions and also in T-cell/B-cell interaction during early lymphoid development.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.3 kDa. The protein migrates as 26-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_006128.1 | |
Human CD7, His Tag (CD7-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1). | |
-20°C | |
CD7 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
18.3 kDa | |
Ala 26 - Pro 180 | |
CD7,GP40,TP41,LEU-9,Tp40 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |