Learn More
ACROBiosystems Human CD99 / MIC2 Protein, Fc Tag
Marke: ACROBiosystems CD9-H5257-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
CD99 antigen is also known as MIC2, single-chain type-1 glycoprotein, HBA71, MIC2X, MIC2Y, MSK5X, 12E7, which belongs to the CD99 family. CD99 / MIC2 is expressed on all leukocytes but highest on thymocytes. Involved in T-cell adhesion processes and in spontaneous rosette formation with erythrocytes. CD99 / MIC2 plays a role in a late step of leukocyte extravasation helping leukocytes to overcome the endothelial basement membrane. CD99 acts at the same site as, but independently of PECAM1. There is also experimental evidence that it binds to cyclophilin A.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 36.2 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH02584 | |
1 mg | |
Asp 23 - Asp 122 | |
CD99,HBA71,MIC2,MIC2X,MIC2Y,MSK5X,12E7 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
36.2 kDa | |
Human CD99, Fc Tag (CD9-H5257) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession # AAH02584). | |
-20°C | |
CD99 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |