missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CDC42SE2 Partial ORF (NP_064625, 1 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00056990-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MSEFWLCFNCCIAEQPQPKRRRRIDRSMIGEPTNFVHTAHVGSGDLFSGMNSVSSIQNQMQSKGGYGGGMPANVQMQLVDTKAGSpezifikation
NP_064625 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.98kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSEFWLCFNCCIAEQPQPKRRRRIDRSMIGEPTNFVHTAHVGSGDLFSGMNSVSSIQNQMQSKGGYGGGMPANVQMQLVDTKAG | |
RUO | |
CDC42SE2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56990 | |
CDC42SE2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ21967/SPEC2 | |
CDC42SE2 | |
Recombinant | |
wheat germ expression system |