Learn More
ACROBiosystems Human Chitinase 3-like 1 / CHI3L1 Protein, His Tag
Marke: ACROBiosystems CH1-H5228-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Chitinase-3-like protein 1 (CHI3L1) is also known as Cartilage glycoprotein 39 (CGP-39), YKL-40, which belongs to the glycosyl hydrolase 18 family. CHI3L1 is present in activated macrophages, articular chondrocytes, synovial cells as well as in liver. CHI3L1 may play a role in defense against pathogens, or in tissue remodeling and also may play an important role in the capacity of cells to respond to and cope with changes in their environment.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.2 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH38354 | |
1 mg | |
Tyr 22 - Thr 383 | |
CHI3L1,YKL-40,CGP-39,GP-39,ASRT7 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
41.2 kDa | |
Human CHI3L1, His Tag (CH1-H5228) is expressed from human 293 cells (HEK293). It contains AA Tyr 22 - Thr 383 (Accession # AAH38354). | |
-20°C | |
Chitinase 3-like 1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |