missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CHST5 Partial ORF (NP_036258.1, 310 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00023563-Q01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16184836

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 18-06-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The carbohydrates of glycoconjugates are highly diverse structures with variation in monosaccharide composition, glycosidic linkage positions, and branching of chains. Further diversity is added by the covalent addition of sulfate moieties to particular hydroxyl groups and amino groups of saccharides. The sulfate modifications of glycoproteins can be extensive in amount and frequently occur at high density. They can have a profound effect on the physiochemical properties of the glycoconjugates, at least in part through the addition of negative charge. Carbohydrate sulfation plays a critical role in many biologic processes. CHST5 belongs to the GST family of sulfotransferases, which also includes CHST1 (MIM 603797), CHST2 (MIM 603798), CHST3 (MIM 603799), and LSST. These enzymes are 6-O-sulfotransferases, which add sulfate to C6 of galactose (Gal), N-acetylgalactosamine (GalNAc), or N-acetylglucosamine (GlcNAc) (Lee et al., 1999 [PubMed 10491328]).[supplied by OMIM]

Sequence: GSGIGKPIEAFHTSSRNARNVSQAWRHALPFTKILRVQEVCAGALQLLGYRPVYSADQQRDLTLDLVLPRGPDHFSWASPD
Spezifikation

Spezifikation

NP_036258.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.65kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GSGIGKPIEAFHTSSRNARNVSQAWRHALPFTKILRVQEVCAGALQLLGYRPVYSADQQRDLTLDLVLPRGPDHFSWASPD
RUO
CHST5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23563
CHST5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ22167/I-GlcNAc-6-ST/MGC74625
CHST5
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote