Learn More
Abnova™ Human CIP29 Full-length ORF (NP_149073.1, 1 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084324-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein that is upregulated in response to various cytokines. The encoded protein may play a role in cell cycle progression. A translocation between this gene and the myeloid/lymphoid leukemia gene, resulting in expression of a chimeric protein, has been associated with acute myelomonocytic leukemia. Pseudogenes exist on chromosomes 7 and 8. Alternatively spliced transcript variants have been described. [provided by RefSeq]
Sequence: MATETVELHKLKLAELKQECLARGLETKGIKQDLIHRLQAYLEEHAEEEANEEDVLGDETEEEETKPIELPVKEEEPPEKTVDVAAEKKVVKITSEIPQTERMQKRAERFNVPVSLESKKAARAARFGISSVPTKGLSSDNKPMVNLDKLKERAQRFGLNVSSISRKSEDDEKLKKRKERFGIVTSSAGTGTTEDTEAKKRKRAERFGIASpezifikation
NP_149073.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HCC-1/HCC1/HSPC316/MGC14726 | |
CIP29 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84324 | |
CIP29 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATETVELHKLKLAELKQECLARGLETKGIKQDLIHRLQAYLEEHAEEEANEEDVLGDETEEEETKPIELPVKEEEPPEKTVDVAAEKKVVKITSEIPQTERMQKRAERFNVPVSLESKKAARAARFGISSVPTKGLSSDNKPMVNLDKLKERAQRFGLNVSSISRKSEDDEKLKKRKERFGIVTSSAGTGTTEDTEAKKRKRAERFGIA | |
RUO | |
CIP29 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |