Learn More
ACROBiosystems Human COMP / TSP5 Protein, His Tag
302.42 € - 2020.12 €
Spezifikation
Zugriffsnummer | AAI25093 |
---|---|
Molekulargewicht | 81.9 kDa |
Quelle | Human COMP, His Tag (COP-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Ala 757 (Accession # AAI25093). |
Immunogen | Gln 21 - Ala 757 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15826467
|
ACROBiosystems
COP-H5220-100UG |
100 μg |
302.42 €
100 Mikrogramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15836467
|
ACROBiosystems
COP-H5220-1MG |
1 mg |
2020.12 €
1 Milligramm |
Verfügbar ab: 28-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Cartilage oligomeric matrix protein (COMP) is also known as Thrombospondin-5 (TSP5), EDM1, EPD1, MED, PSACH, THBS5, which belongs to the thrombospondin family. COMP / TSP5 contains 4 EGF-like domains, 1 TSP C-terminal (TSPC) domain, 8 TSP type-3 repeats. Abundantly expressed in the chondrocyte extracellular matrix, and is also found in bone, tendon, ligament and synovium and blood vessels. COMP may play a role in the structural integrity of cartilage via its interaction with other extracellular matrix proteins such as the collagens and fibronectin. COMP can mediate the interaction of chondrocytes with the cartilage extracellular matrix through interaction with cell surface integrin receptors. Thrombospondin-5 could play a role in the pathogenesis of osteoarthritis. COMP is a marker of cartilage turnover.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 81.9 kDa. The protein migrates as 110-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAI25093 | |
Human COMP, His Tag (COP-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Ala 757 (Accession # AAI25093). | |
-20°C | |
COMP | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
81.9 kDa | |
Gln 21 - Ala 757 | |
COMP,TSP5,EDM1,EPD1,MED,PSACH,THBS5 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>88% as determined by SDS-PAGE |