Learn More
Abnova™ Human CYTL1 Partial ORF (NP_061129, 37 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00054360-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
C17 is a cytokine-like protein specifically expressed in bone marrow and cord blood mononuclear cells that bear the CD34 (MIM 142230) surface marker (Liu et al., 2000 [PubMed 10857752]).[supplied by OMIM]
Sequence: EITRDFNLLQVSEPSEPCVRYLPRLYLDIHNYCVLDKLRDFVASPPCWKVAQVDSLKDKARKLYTIMNSFCRRDLVFLLDDCNALEYPIPVTTVLPDRQRSpezifikation
NP_061129 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EITRDFNLLQVSEPSEPCVRYLPRLYLDIHNYCVLDKLRDFVASPPCWKVAQVDSLKDKARKLYTIMNSFCRRDLVFLLDDCNALEYPIPVTTVLPDRQR | |
RUO | |
CYTL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54360 | |
CYTL1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C17/C4orf4 | |
CYTL1 | |
Recombinant | |
wheat germ expression system |