Learn More
Abnova™ Human DGCR14 Partial ORF (NP_073210, 377 a.a. - 476 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00008220-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is located within the minimal DGS critical region (MDGCR) thought to contain the gene(s) responsible for a group of developmental disorders. These disorders include DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome, and some familial or sporadic conotruncal cardiac defects which have been associated with microdeletion of 22q11.2. The encoded protein may be a component of C complex spliceosomes, and the orthologous protein in the mouse localizes to the nucleus. [provided by RefSeq]
Sequence: RVTENLASLTPKGLSPAMSPALQRLVSRTASKYTDRALRASYTPSPARSTHLKTPASGLQTPTSTPAPGSATRTPLTQDPASITDNLLQLPARRKASDFFSpezifikation
NP_073210 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RVTENLASLTPKGLSPAMSPALQRLVSRTASKYTDRALRASYTPSPARSTHLKTPASGLQTPTSTPAPGSATRTPLTQDPASITDNLLQLPARRKASDFF | |
RUO | |
DGCR14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8220 | |
DGCR14 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DGCR13/DGS-H/DGS-I/DGSH/DGSI/ES2/Es2el | |
DGCR14 | |
Recombinant | |
wheat germ expression system |