Learn More
ACROBiosystems Human DPPIV / CD26 Protein, Tag Free (Active dimer) (active enzyme)
Marke: ACROBiosystems DP4-H5211-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Dipeptidyl peptidase-IV (DPPIV) is also known as adenosine deaminase complexing protein 2, DPPIV or CD26 is antigenic enzyme expressed on the surface of most cell types and is associated with immune regulation, signal transduction and apoptosis. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. The substrates of DPPIV are proline (or alanine)-containing peptides and include growth factors, chemokines, neuropeptides, and vasoactive peptides. DPPIV plays a major role in glucose metabolism. It is responsible for the degradation of incretins such as GLP-1. DPPIV plays an important role in tumor biology, and is useful as a marker for various cancers, with its levels either on the cell surface or in the serum increased in some neoplasms and decreased in others. DPPIV also binds the enzyme adenosine deaminase specifically and with high affinity. The significance of this interaction has yet to be established.
This protein carries no "e;tag"e;. The protein has a calculated MW of 85.5 kDa. The protein migrates as 90-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001926.2 | |
50 μg | |
Asn 29 - Pro 766 | |
DPP4,ADABP,ADCP2,CD26,DPPIV,TP103 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
85.5 kDa | |
Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession # NP_001926.2). | |
-20°C | |
DPPIV | |
Human | |
50 mM HEPES, 150 mM NaCl, pH7.5 | |
HEK293 | |
Enzymes |