missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EMP3 Partial ORF (NP_001416.1, 25 a.a. - 68 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002014-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: DKSWWTLPGKESLNLWYDCTWNNDTKTWACSNVSENGWLKAVQVSpezifikation
NP_001416.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
30.58kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DKSWWTLPGKESLNLWYDCTWNNDTKTWACSNVSENGWLKAVQV | |
RUO | |
EMP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2014 | |
EMP3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
YMP | |
EMP3 | |
Recombinant | |
wheat germ expression system |