Learn More
ACROBiosystems Human FABP1 / L-FABP Protein, His Tag
Marke: ACROBiosystems FA1-H5148-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Fatty acid-binding protein 1 (FABP1) is also known as Fatty acid-binding protein, liver (FABPL), Liver-type fatty acid-binding protein (L-FABP). FABP1 / FABPL belongs to the calycin superfamily and Fatty-acid binding protein (FABP) family. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 / L-FABP binds free fatty acids and their coenzyme A derivatives, bilirubin, and some other small molecules in the cytoplasm. FABP-1 may be involved in intracellular lipid transport.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.0 kDa. The protein migrates as 15-16 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_001434 | |
1 mg | |
Ser 2 - Ile 127 | |
FABP1,FABPL,L-FABP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
15.0 kDa | |
Human FABP1, His Tag (FA1-H5148) is expressed from E.coli cells. It contains AA Ser 2 - Ile 127 (Accession # NP_001434). | |
-20°C | |
FABP1 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Bio-Markers and CD Antigens |