Learn More
Abnova™ Human FBXO43 Partial ORF (XP_209918, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00286151-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Members of the F-box protein family, such as FBXO43, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MSQRHSGQAGTEAGNGADSPPIVNSKYSTFRDFCSTSSFQDSGYNELKSCSFDNIDKEYLGKKEKGPTLLYEHPETSGLGLTHPLESPTQKKKCILPRKEKDKTPELCETSpezifikation
XP_209918 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSQRHSGQAGTEAGNGADSPPIVNSKYSTFRDFCSTSSFQDSGYNELKSCSFDNIDKEYLGKKEKGPTLLYEHPETSGLGLTHPLESPTQKKKCILPRKEKDKTPELCET | |
RUO | |
FBXO43 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
286151 | |
FBXO43 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EMI2/ERP1/Fbx43 | |
FBXO43 | |
Recombinant | |
wheat germ expression system |