missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RI/CD64 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF1257
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Polyclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| Fc gamma RI/CD64 | |
| Polyclonal | |
| Unconjugated | |
| P12314.2 | |
| FCGR1A | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RI/CD64 Gln16-Pro288 Accession # P12314.2 | |
| 100 μg | |
| Primary | |
| Detects human Fc gamma RI/CD64 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Fc gamma RI is observed and 10% cross-reactivity with recombinant human (rh) Fc gamma RIIA and rhFc gamma RIIIB is observed. | |
| Human | |
| IgG |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2209 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction