Learn More
ACROBiosystems Human Frizzled-2 / FZD2 (24-190) Protein, Fc Tag
Marke: ACROBiosystems FZ2-H5255-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Frizzled-2 (FZD2) is also known as FzE2, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD2 contains one FZ (frizzled) domain. FZD2 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. The Lys-Thr-X-X-X-Trp motif of FZD2 interacts with the PDZ doman of Dvl (Disheveled) family members and is involved in the activation of the Wnt/beta-catenin signaling pathway.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.4 kDa. The protein migrates as 50-62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001457.1 | |
100 μg | |
Gln 24 - Pro 190 | |
FZD2,Frizzled-2,Fz-2,hFz2,FzE2 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
44.4 kDa | |
Human Frizzled-2, Fc Tag (FZ2-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Pro 190 (Accession # Q14332-1). | |
-20°C | |
Frizzled-2 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |