Learn More
ACROBiosystems Human Frizzled-5 / FZD5 Protein, Fc Tag
Marke: ACROBiosystems FZ5-H5259-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Frizzled-5 (FZD5) is also known as FzE5, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD5 contains one FZ (frizzled) domain. FZD5 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. FZD5 interacts specifically with Wnt5A to induce the beta-catenin pathway. FZD5 interacts with GOPC.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
Q13467 | |
1 mg | |
Ala 27 - Pro 167 | |
FZD5,Frizzled-5,C2orf31,,Fz-5,hFz5,FzE5 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
42.6 kDa | |
Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Pro 167 (Accession # Q13467). | |
-20°C | |
Frizzled-5 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |