missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Frizzled-7 / FZD7 Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | NP_003498.1 |
---|---|
Molekulargewicht | 18.6 kDa |
Quelle | Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 33 - Leu 185 (Accession # O75084-1). |
Immunogen | Gln 33 - Leu 185 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16417569
|
ACROBiosystems
FZ7-H52H9-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16427569
|
ACROBiosystems
FZ7-H52H9-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Frizzled-7 (FZD7) is also known as FzE3, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD7 contains one FZ (frizzled) domain. FZD7 is receptor for Wnt proteins. FZD7 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. FZD7 interacts with MAGI3 and DVL1.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_003498.1 | |
Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 33 - Leu 185 (Accession # O75084-1). | |
-20°C | |
Frizzled-7 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
18.6 kDa | |
Gln 33 - Leu 185 | |
FZD7,Frizzled-7,FzE3,Fz-7,hFz7 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |