Learn More
Abnova™ Human GNAT2 Full-length ORF (NP_005263.1, 1 a.a. - 354 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002780-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in cones. [provided by RefSeq]
Sequence: MGSGASAEDKELAKRSKELEKKLQEDADKEAKTVKLLLLGAGESGKSTIVKQMKIIHQDGYSPEECLEFKAIIYGNVLQSILAIIRAMTTLGIDYAEPSCADDGRQLNNLADSIEEGTMPPELVEVIRRLWKDGGVQACFERAAEYQLNDSASYYLNQLERITDPEYLPSEQDVLRSRVKTTGIIETKFSVKDLNFRMFDVGGQRSERKKWIHCFEGVTCIIFCAALSAYDMVLVEDDEVNRMHESLHLFNSICNHKFFAATSIVLFLNKKDLFEEKIKKVHLSICFPEYDGNNSYDDAGNYIKSQFLDLNMRKDVKEIYSHMTCATDTQNVKFVFDAVTDIIIKENLKDCGLFSpezifikation
NP_005263.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACHM4/GNATC | |
GNAT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2780 | |
GNAT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGSGASAEDKELAKRSKELEKKLQEDADKEAKTVKLLLLGAGESGKSTIVKQMKIIHQDGYSPEECLEFKAIIYGNVLQSILAIIRAMTTLGIDYAEPSCADDGRQLNNLADSIEEGTMPPELVEVIRRLWKDGGVQACFERAAEYQLNDSASYYLNQLERITDPEYLPSEQDVLRSRVKTTGIIETKFSVKDLNFRMFDVGGQRSERKKWIHCFEGVTCIIFCAALSAYDMVLVEDDEVNRMHESLHLFNSICNHKFFAATSIVLFLNKKDLFEEKIKKVHLSICFPEYDGNNSYDDAGNYIKSQFLDLNMRKDVKEIYSHMTCATDTQNVKFVFDAVTDIIIKENLKDCGLF | |
RUO | |
GNAT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |