missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GPR119 Partial ORF (NP_848566.1, 236 a.a. - 335 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00139760-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
GPR119 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) (Fredriksson et al., 2003 [PubMed 14623098]).[supplied by OMIM]
Sequence: LSWTPFLITGIVQVACQECHLYLVLERYLWLLGVGNSLLNPLIYAYWQKEVRLQLYHMALGVKKVLTSFLLFLSARNCGPERPRESSCHIVTISSSEFDGSpezifikation
NP_848566.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
LSWTPFLITGIVQVACQECHLYLVLERYLWLLGVGNSLLNPLIYAYWQKEVRLQLYHMALGVKKVLTSFLLFLSARNCGPERPRESSCHIVTISSSEFDG | |
RUO | |
GPR119 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
139760 | |
GPR119 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPCR2/MGC119957/hGPCR2 | |
GPR119 | |
Recombinant | |
wheat germ expression system |