Learn More
ACROBiosystems Human HABP1 / C1QBP Protein, His Tag
Marke: ACROBiosystems C1P-H5122-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Complement component 1 Q subcomponent-binding protein, mitochondrial (C1QBP), a member of the MAM33 family, is also known as ASF/SF2-associated protein p32, glycoprotein gC1qBP (C1qBP), mitochondrial matrix protein p32 and hyaluronan-binding protein 1, which is a homotrimer that contains three monomers forming a donut-shaped structure with an unusually asymmetric charge distribution on the surface. C1q associates with C1r and C1s in order to yield the first component of the serum complement system. C1QBP has also been identified as the p32 subunit of pre-mRNA splicing factor SF2, as well as a hyaluronic acid-binding protein. Furthermore, C1QBP has been shown to interact with Protein kinase D1, BAT2, PRKCD, PKC alpha and Protein kinase Mζ .
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.6 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_001203.1 | |
100 μg | |
His 75 - Gln 282 | |
C1QBP,GC1QBP,HABP1,SF2P32,p33 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
24.6 kDa | |
Human HABP1, His Tag (C1P-H5122) is expressed from E.coli cells. It contains AA His 75 - Gln 282 (Accession # Q07021-1). | |
-20°C | |
HABP1 | |
Human | |
PBS, pH7.4 | |
E.coli | |
Other Recombinant Proteins |