Learn More
ACROBiosystems Human Her2 / ErbB2 Protein, Tag Free
411.28 € - 1693.52 €
Spezifikation
Zugriffsnummer | NP_004439.2 |
---|---|
Molekulargewicht | 70.1 kDa |
Quelle | Human Her2, Tag Free (HE2-H5212) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1). |
Immunogen | Thr 23 - Thr 652 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16497619
|
ACROBiosystems
HE2-H5212-50UG |
50 μg |
411.28 €
50 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16487619
|
ACROBiosystems
HE2-H5212-500UG |
500 μg |
1693.52 €
500 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Human Epidermal growth factor Receptor 2 (HER2) is also called ERBB2, HER-2,HER-2 /neu, NEU, NGL,TKR1 and c-erb B2,and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family, more commonly known as the epidermal growth factor receptor family. HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. HER2 is thought to be an orphan receptor, with none of the EGF family of ligands able to activate it. Approximately 30% of breast cancers have an amplification of the HER2 gene or overexpression of its protein product. Overexpression of this receptor in breast cancer is associated with increased disease recurrence and worse prognosis. HER2 appears to play roles in development, cancer, communication at the neuromuscular junction and regulation of cell growth and differentiation .
This protein carries no "e;tag"e;. The protein has a calculated MW of 70.1 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_004439.2 | |
Human Her2, Tag Free (HE2-H5212) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1). | |
-20°C | |
Her2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
70.1 kDa | |
Thr 23 - Thr 652 | |
ERBB2,CD340,HER-2/neu,HER2,MLN19,NEU,NGL,TKR1 | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |