Learn More
Abnova™ Human HOMER2 Full-length ORF (NP_004830.2, 1 a.a. - 343 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009455-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function. The encoded protein may be involved in cell growth. Four transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MGEQPIFTTRAHVFQIDPNTKKNWMPASKQAVTVSYFYDVTRNSYRIISVDGAKVIINSTITPNMTFTKTSQKFGQWADSRANTVFGLGFSSEQQLTKFAEKFQEVKEAAKIAKDKTQEKIETSSNHSQASSVNGTDDEKASHAGPANTHLKSENDKLKIALTQSAANVKKWEIELQTLRESNARLTTALQESAASVEQWKRQFSICRDENDRLRNKIDELEEQCSEINREKEKNTQLKRRIEELEAELREKETELKDLRKQSEIIPQLMSECEYVSEKLEAAERDNQNLEDKVRSLKTDIEESKYRQRHLKVELKSFLEVLDGKIDDLHDFRRGLSKLGTDNSpezifikation
NP_004830.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACPD/CPD/HOMER-2/HOMER2A/HOMER2B/Vesl-2 | |
HOMER2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9455 | |
HOMER2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGEQPIFTTRAHVFQIDPNTKKNWMPASKQAVTVSYFYDVTRNSYRIISVDGAKVIINSTITPNMTFTKTSQKFGQWADSRANTVFGLGFSSEQQLTKFAEKFQEVKEAAKIAKDKTQEKIETSSNHSQASSVNGTDDEKASHAGPANTHLKSENDKLKIALTQSAANVKKWEIELQTLRESNARLTTALQESAASVEQWKRQFSICRDENDRLRNKIDELEEQCSEINREKEKNTQLKRRIEELEAELREKETELKDLRKQSEIIPQLMSECEYVSEKLEAAERDNQNLEDKVRSLKTDIEESKYRQRHLKVELKSFLEVLDGKIDDLHDFRRGLSKLGTDN | |
RUO | |
HOMER2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |