missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ICAT Antibody, R&D Systems™

Goat Polyclonal Antibody
Marke: R&D Systems AF3016-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ICAT/CTNNBIP1 Polyclonal specifically detects ICAT/CTNNBIP1 in Human samples. It is validated for Western Blot.
Spezifikation
| ICAT/CTNNBIP1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-catenin-interacting protein ICAT, catenin, beta interacting protein 1, CTNNBIP1, ICATbeta-interacting protein 1, Inhibitor of beta-catenin and Tcf-4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 56998 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9NSA3 | |
| CTNNBIP1 | |
| E. coli-derived recombinant human ICAT Met1-Gln81 Accession # Q9NSA3 | |
| 25 μg | |
| Primary | |
| Detects human ICAT in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur