Learn More
ACROBiosystems Human IL-21 R Protein, His Tag
Marke: ACROBiosystems ILR-H5226-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Interleukin-21 receptor (IL21R) is also known as CD antigen CD360, Novel interleukin receptor (NILR), is a receptor for interleukin-21 and belongs to the type I cytokine receptor family or type 4 subfamily. IL21R contains two fibronectin type-III domains. IL21R is selectively expressed in lymphoid tissues and most highly expressed in thymus and spleen. IL21R transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.1 kDa. The protein migrates as 36-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH04348 | |
1 mg | |
Cys 20 - Pro 236 | |
IL21R,CD360,NILR | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
26.1 kDa | |
Human IL-21 R, His Tag (ILR-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348). | |
-20°C | |
IL-21 R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |