missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-24 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 583.00 €
Spezifikation
| Antigen | IL-24 |
|---|---|
| Klon | 283161 |
| Konzentration | LYOPH |
| Verdünnung | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Anwendungen | ELISA |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18730197
![]() |
R&D Systems
MAB19652-SP |
25 μg |
209.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18190181
|
R&D Systems
MAB19652 |
500 μg |
583.00 €
500 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
IL-24 Monoclonal specifically detects IL-24 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Spezifikation
| IL-24 | |
| LYOPH | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q13007 | |
| 11009 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-24 Gln50-Leu206 Accession # Q13007 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-24 in ELISAs. |
| 283161 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C49A, FISP, IL10B, IL24, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, MOB-5, ST16, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein | |
| IL24 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts