Learn More
ACROBiosystems Human IL-33 Protein, His Tag
423.38 € - 3495.90 €
Spezifikation
Zugriffsnummer | AAH47085.1 |
---|---|
Molekulargewicht | 20.2 kDa |
Quelle | Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession # O95760-1). |
Immunogen | His 109 - Thr 270 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16467699
|
ACROBiosystems
IL3-H52H7-50UG |
50 μg |
423.38 €
50 Mikrogramm |
Verfügbar ab: 17-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16457699
|
ACROBiosystems
IL3-H52H7-1MG |
1 mg |
3495.90 €
1 Milligramm |
Verfügbar ab: 17-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Interleukin 33 (IL33) is known as C9orf26, DKFZp586H0523, DVS27, NF-HEV, NFEHEV, RP11-575C20.2,and is a cytokine belonging to the IL-1 superfamily. IL-33 induces helper T cells, mast cells, eosinophils and basophils to produce type 2 cytokines. IL-33 mediates its biological effects by interacting with the receptors ST2 (aka IL1RL1) and IL-1 Receptor Accessory Protein (IL1RAP), activating intracellular molecules in the NF-κB and MAP kinase signaling pathways that drive production of type 2 cytokines (e.g. IL-5 and IL-13) from polarized Th2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH47085.1 | |
Human IL-33, His Tag (IL3-H52H7) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession # O95760-1). | |
-20°C | |
IL-33 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |
20.2 kDa | |
His 109 - Thr 270 | |
IL33,DV27,C9ORF26,IL1F11,NFHEV,DKFZp586H0523,DVS27,NFEHEV,RP11-575C20.2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |