Learn More
ACROBiosystems Human ILDR2 Protein, Fc Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | NP_955383.1 |
---|---|
Molekulargewicht | 45.1 kDa |
Quelle | Human ILDR2, Fc Tag (IL2-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Glu 186 (Accession # Q71H61-1). |
Immunogen | Leu 21 - Glu 186 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17136401
|
ACROBiosystems
IL2-H5259-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
17146401
|
ACROBiosystems
IL2-H5259-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
ILDR2 belongs to the immunoglobulin superfamily. May be involved in lipid homeostasis and ER stress pathways. ILDR2 as a novel B7-like protein with robust T cell inhibitory activity, expressed in immune cells and in immune-privileged and inflamed tissues. A fusion protein, consisting of ILDR2 extracellular domain with an Fc fragment, that binds to a putative counterpart on activated T cells showed a beneficial effect in the collagen-induced arthritis model and abrogated the production of proinflammatory cytokines and chemokines in autologous synovial-like cocultures of macrophages and cytokine-stimulated T cells. Collectively, these findings point to ILDR2 as a novel negative regulator for T cells, with potential roles in the development of immune-related diseases, including autoimmunity and cancer.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.1 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Spezifikation
NP_955383.1 | |
Human ILDR2, Fc Tag (IL2-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Glu 186 (Accession # Q71H61-1). | |
-20°C | |
ILDR2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
45.1 kDa | |
Leu 21 - Glu 186 | |
ILDR2,C1orf32 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |