Learn More
ACROBiosystems Human Insulin R / CD220 (28-944) Protein, His Tag
SPR verified
387.09 € - 2794.30 €
Spezifikation
Zugriffsnummer | NP_000199.2 |
---|---|
Molekulargewicht | 106.5 kDa |
Quelle | Human Insulin R (28-944), His Tag (SPR verified) (INR-H52Ha) is expressed from human 293 cells (HEK293). It contains AA His 28 - Lys 944 (Accession # P06213-2). |
Immunogen | His 28 - Lys 944 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16467779
|
ACROBiosystems
INR-H52HA-1MG |
1 mg |
2794.30 €
1 Milligramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16457779
|
ACROBiosystems
INR-H52HA-100UG |
100 μg |
387.09 €
100 Mikrogramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Insulin receptor (INSR) is also known as CD antigen CD220, which can be cleaved into the following 2 chains: Insulin receptor subunit alpha and Insulin receptor subunit beta. INSR is a tetramer of 2 alpha and 2 beta chains linked by disulfide bonds. The alpha chains carry the insulin-binding regions, while the beta chains carry the kinase domain. Forms a hybrid receptor with IGF1R, the hybrid is a tetramer consisting of 1 alpha chain and 1 beta chain of INSR and 1 alpha chain and 1 beta chain of IGF1R. In addition to binding insulin, the insulin receptor can bind insulin-like growth factors (IGFI and IGFII). Isoform Short of INSR has a higher affinity for IGFII binding. When present in a hybrid receptor with IGF1R, INSR binds IGF1.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 106.5 kDa. The protein migrates as 115-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000199.2 | |
Human Insulin R (28-944), His Tag (SPR verified) (INR-H52Ha) is expressed from human 293 cells (HEK293). It contains AA His 28 - Lys 944 (Accession # P06213-2). | |
-20°C | |
Insulin R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
106.5 kDa | |
His 28 - Lys 944 | |
INSR,Insulin receptor,IR,CD220 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |